Isatin for synthesis, Sigma-Aldrich®
: Merck
:91-56-5
:
:C₈H₅NO₂
:MFCD00005718
:147.13 g/mol
:13,05119
:342 °C (1013 hPa)
:198…202 °C
:1,472 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:220 °C
:Ambient
:202-077-8
- Solubility:2 g/l (20 °C)
- Melting Point:199 - 204 °C
- Molar Mass:147.13 g/mol
- Bulk Density:600 kg/m3
- Flash Point:200 °C
- pH:7 (1.9 g/l, H2O, 20 °C)
- Ignition Point:220 °C
- Hill Formula:C8H5NO2
Frequently Bought Together