
2-Methylbenzimidazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-15-6
:
Formula:C₈H₈N₂
MDL Number:MFCD00005598
:132.16 g/mol
Boiling Pt:339 °C (1013 hPa)
Melting Pt:174…178 °C
Storage Temperature:Ambient
EINECS:210-411-9
:GHS07
Frequently Bought Together