Benzimidazole
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:51-17-2
:
Formula:C₇H₆N₂
MDL Number:MFCD00005585
:118.14 g/mol
Merck Index:13,01082
Boiling Pt:360 °C (1013 hPa)
Melting Pt:170…173 °C
Density:1,23 g/cm³ (20 °C)
EINECS:200-081-4
Frequently Bought Together