Order Entry
Northern Ireland
ContactUsLinkComponent
4-Methylpiperidine for synthesis, Sigma-Aldrich®
Flammable
Corrosive
4-Methylpiperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
4-Methylpiperidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    4 - 5 °C
  • Molar Mass:
    99.18 g/mol
  • Boiling Point:
    121 - 127 °C
  • Vapor Pressure:
    16 hPa (20 °C)
  • Flash Point:
    13 °C
  • Refractive Index:
    1.4487 (20 °C, 589 nm)
  • Density:
    0.84 g/cm3 (20 °C)
  • pH:
    13 (100 g/l, H2O, 20 °C)
  • Synonyms:
    4-Pipecoline
  • Hill Formula:
    C6H13N

Specification Test Results

Related Information

Frequently Bought Together