N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
: Merck
:6066-82-6
:
:C₄H₅NO₃
:MFCD00005516
:115.09 g/mol
:363 °C (1013 hPa)
:93…95 °C
:M-Clarity™ quality level = MQ200
:Refrigerator
:228-001-3
- Solubility:(20 °C) soluble
- Melting Point:93 - 100 °C
- Molar Mass:115.09 g/mol
- pH:5 - 7 (50 g/l, H2O, 20 °C)
Frequently Bought Together