Order Entry
Northern Ireland
ContactUsLinkComponent
2-Fluorocinnamic acid predominantly trans 98%
2-Fluorocinnamic acid predominantly trans 98%
Catalog # A15303.14
CAS Number:  
2-Fluorocinnamic acid predominantly trans 98%
Catalog # A15303.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

CAS Number:451-69-4
 :
Formula:C₉H₇FO₂
MDL Number:MFCD00004370
 :166.15 g/mol
Melting Pt:179…183 °C
EINECS:207-195-3
 :GHS07
Storage Temperature:Ambient