
trans-2-Hydroxycinnamic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:614-60-8
:
Formula:C₉H₈O₃
MDL Number:MFCD00004379
:164.16 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:∼ 210 °C (dec.)
Storage Temperature:Ambient
EINECS:210-386-4
:GHS07
Related Information
Frequently Bought Together