
2,5-Dimethoxycinnamic acid predominantly trans 99%
Catalog # B21649.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:10538-51-9
:
Formula:C₁₁H₁₂O₄
MDL Number:MFCD00004378
:208.21 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:147…150 °C
Storage Temperature:Ambient
EINECS:234-114-9
:GHS07
:38489-74-6