
trans-2-Methoxycinnamic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1011-54-7
:6099-03-2
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00064238
:178.19 g/mol
Melting Pt:183…186 °C
EINECS:213-785-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together