Order Entry
Northern Ireland
ContactUsLinkComponent
4'-Nitrocinnamic acid for synthesis
4'-Nitrocinnamic acid for synthesis
Catalog # 8.18563.0025
Supplier:  Merck
CAS Number:  
4'-Nitrocinnamic acid for synthesis
Catalog # 8.18563.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    288 - 291 °C
  • Molar Mass:
    193.15 g/mol
  • Synonyms:
    trans-3-(4-Nitrophenyl)propenoic acid
  • Hill Formula:
    C9H7NO4
  • Chemical Formula:
    4-(NO2)C6H4CH=CHCOOH

Specification Test Results

About this item

 :
 :193.16 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:212-924-3
CAS Number:882-06-4
 :619-89-6
Formula:C₉H₇NO₄
MDL Number:MFCD00007381