
cis-4-Hexen-1-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:928-91-6
:
Formula:C₆H₁₂O
MDL Number:MFCD00148974
Boiling Pt:158 °C
Density:0,857
Flash Pt:65 °C (149 °F)
Storage Temperature:Ambient
EINECS:213-187-0
UN:1987
ADR:3,III
:100.16 g/mol
:GHS02
Related Information
Frequently Bought Together