

cis-2-Hexen-1-ol ≥94% remainder mainly trans isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:928-94-9
:
Formula:C₆H₁₂O
MDL Number:MFCD00063209
:100.16 g/mol
Boiling Pt:158 °C
Density:0,847
Flash Pt:54 °C (129 °F)
Storage Temperature:Ambient
EINECS:213-190-7
UN:1987
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together