1,1'-[Oxybis(ethyleneoxy)]diethylene for synthesis, Sigma-Aldrich®
Catalog # 8.40144.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 ml
- Seq:2370
Specifications
Specification Test Results
- Molar Mass:158.20 g/mol
- Density:0.97 g/cm3
Specification Test Results
About this item
:
Density:0,97 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:158.2 g/mol
EINECS:212-133-3
CAS Number:764-99-8
Formula:H₂CCHO(CH₂)₂O(CH₂)₂OCHCH₂
MDL Number:MFCD00054610