Order Entry
Northern Ireland
ContactUsLinkComponent
 
Toxic
1,1,2-Trichloro-3,3,3-trifluoropropene ≥97%
Catalog # L09263.14
CAS Number:  
Toxic
1,1,2-Trichloro-3,3,3-trifluoropropene ≥97%
Catalog # L09263.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :199.39 g/mol
Boiling Pt:88 °C
Melting Pt:–114 °C
Density:1,617
Storage Temperature:Ambient
EINECS:207-075-0
UN:2810
ADR:6.1,III
 :GHS06
CAS Number:431-52-7
Formula:C₃Cl₃F₃
MDL Number:MFCD00013672