
(4-Hydroxyphenyl)acetic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156-38-7
:
Formula:(C₆H₄(OH))CH₂COOH
MDL Number:MFCD00004347
:152.15 g/mol
Boiling Pt:347 °C (1013 hPa)
Melting Pt:147…152 °C
Storage Temperature:Ambient
EINECS:205-851-3
:GHS07
Specification Test Results
- Appearance:White to cream to tan Crystalline powder
- Infrared spectrum:Conforms
- Melting point:147 to 152 °C
- Water:≤0.5 % (K.F.)
- TLC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together