Order Entry
Northern Ireland
ContactUsLinkComponent
(4-Chlorophenyl)acetic acid methyl ester 99%
(4-Chlorophenyl)acetic acid methyl ester 99%
Certificates
undefined
(4-Chlorophenyl)acetic acid methyl ester 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5220 to 1.5240 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together