
(4-Chlorophenyl)acetic acid ethyl ester 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14062-24-9
:
Formula:(C₆H₄Cl)CH₂COOCH₂CH₃
MDL Number:MFCD00018791
:198.65 g/mol
EINECS:237-906-2
:GHS07
Specification Test Results
- Appearance:Colorless Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together