Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

(2,6-Dichlorophenyl)acetic acid methyl ester 99%

Irritant
(2,6-Dichlorophenyl)acetic acid methyl ester 99%
Certificates
(2,6-Dichlorophenyl)acetic acid methyl ester 99%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Methyl 2-(2,6-dichlorophenyl)acetate, Methyl 2,6-Dichlorophenylacetate, 2,6-Dichlorophenylacetic acid methyll ester, Guanfacine Methyl Ester Impurity, Methyl (2,6-dichlorophenyl)acetate
  • CAS Number:54551-83-6
  • Formula:(C₆H₃Cl₂)CH₂COOCH₃
  • MDL Number:MFCD00191640
  • Molecular Weight:219.07 g/mol
  • EINECS:259-221-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5400 to 1.5420 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £74.50
    £74.50
    1 * 5 g
       £74.50

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £225.00
    £225.00
    1 * 25 g
       £225.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.