Order Entry
Northern Ireland
ContactUsLinkComponent
3-(Indol-3-yl)propionic acid 99%
Irritant
3-(Indol-3-yl)propionic acid 99%
Certificates
undefined
Irritant
3-(Indol-3-yl)propionic acid 99%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    130 to 136 °C
  • Water:
    ≤0.5 % (K.F.)
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together