
1-Naphthylacetic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:86-87-3
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00004046
:186.21 g/mol
Merck Index:12,06458
Boiling Pt:352 °C (1013 hPa)
Melting Pt:129…131 °C
Density:∼1,263 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-705-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.42 g/l (20 °C)
- Melting Point:125 - 131 °C
- Molar Mass:186.21 g/mol
- Bulk Density:240 kg/m3
- pH:3.0 (0.6 g/l, H2O, 25 °C)
Specification Test Results
Frequently Bought Together