Calbiochem® Phaseolus vulgaris Lectin, PHA-L
Catalog # 431784-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:5 mg
- Species:Phaseolus vulgaris
- Storage conditions:–20 °C
- Form:Lyophilised
- Protein synonyms:Red kidney bean isolectin L
- Protein/peptide name:Lectin, PHA-L
- Purity:Homogeneous
Specifications
About this item
PHA-L is a potent lymphocyte mitogen with a narrow range. Requires calcium and manganese ions for activity. It has also been found to be useful as a specific marker in anterograde neuronal tracing.
- Elution: 100 mM acetic acid