


Diisobutylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-96-3
:
Formula:C₈H₁₉N
MDL Number:MFCD00008930
:129.25 g/mol
Boiling Pt:137…139 °C (760 mmHg)
Melting Pt:–77 °C
Density:0,74 g/cm³
Flash Pt:26 °C
Storage Temperature:Ambient
EINECS:203-819-3
UN:2361
ADR:3,III
:GHS05
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4071 to 1.4091 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤15 APHA
- Sulfate (SO4):≤0.1 %
- Arsenic (As):≤3 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.1 %
Specification Test Results
Frequently Bought Together