


tert-Butyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:590-42-1
:
Formula:C₅H₉NS
MDL Number:MFCD00004816
:115.2 g/mol
Boiling Pt:141…145 °C
Melting Pt:10…12 °C
Density:0,908
Flash Pt:38 °C (100 °F)
Storage Temperature:Ambient
EINECS:209-682-6
UN:2929
ADR:6.1,II
:GHS06
:GHS05
:GHS02
Related Information
Frequently Bought Together