


Isobutyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:591-82-2
:
Formula:C₅H₉NS
MDL Number:MFCD00022062
:115.2 g/mol
Boiling Pt:160…162 °C
Density:0,940
Flash Pt:24 °C (75 °F)
EINECS:209-733-2
UN:2924
ADR:3,III
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together