L(-)-Glutathione (reduced form)
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:70-18-8
:
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939
:307.33 g/mol
Merck Index:13,04488
Melting Pt:185…195 °C
Density:1,475 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-725-4
Frequently Bought Together