ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk® Hydrophilic SC-DVB (SO3), SPE Columns
J.T.Baker® BAKERBOND Speedisk® Hydrophilic SC-DVB (SO3), SPE Columns
Catalog # 8111-04
Supplier:  Avantor
undefined
J.T.Baker® BAKERBOND Speedisk® Hydrophilic SC-DVB (SO3), SPE Columns
Catalog # 8111-04
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    3 ml
  • Seq:
    07
  • Bed weight:
    50 mg
  • Phase:
    Hydrophilic SC-DVB
  • Pk:
    1
Specifications
About this item
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology. These resin particles have a large surface area, are highly rigid, are stable over pH range 1 to 14, are water-wettable and are not impacted by sorbent drying. These columns are recommended when advanced detection methods are used.
  • Improved sample preparation recovery
  • Polymer based
General applications: Ionic, basic analytes.
These SPE columns can be used in mixed mode.