
Sodium methanesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2386-57-4
:
Formula:CH₃NaO₃S
MDL Number:MFCD00064389
:118.09 g/mol
EINECS:219-203-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together