Order Entry
Northern Ireland
ContactUsLinkComponent
 
Dithiothreitol (DTT, Cleland's reagent)
Catalog # MOLE19733320-100G
Supplier:  Molekula
CAS Number:  
undefined
Dithiothreitol (DTT, Cleland's reagent)
Catalog # MOLE19733320-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :154.25 g/mol
Boiling Pt:∼125 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:222-468-7
 :GHS07
CAS Number:3483-12-3
 :27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877