Dithiothreitol (DTT, Cleland's reagent) ≥99% for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Boiling Pt:∼125 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:222-468-7
UN:3335
ADR:9,III
:GHS07
Frequently Bought Together