Dithiothreitol (DTT, Cleland's reagent) ≥99.4%, Proteomics Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Boiling Pt:<125 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:>110 °C (open cup)
Storage Temperature:Refrigerator
EINECS:222-468-7
:GHS07
Specification Test Results
- Abs.@283 nm (0.02 M, water):≤0.05
- DNase:none detected
- Electrophoresis (P/F):Pass
- IR:Pass
- Loss on Drying:≤0.5%
- Melting Point:39 – 43°C
- Oxidized DTT:≤0.5%
- Protease:none detected
- Purity (-SH Content):≥99.4%
- RNase:none detected
- Solubility (5%, Water):Pass
Specification Test Results
Frequently Bought Together