Sodium methoxide ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 124-41-4 Molecular Formula: CH3NaO Formula Weight: 54.02 Melting Point: 300° Beilstein Reference: 3592982 Merck Reference: 14,8643 UN No.: 1431 MDL No.: MFCD00012179
Synonyms: Sodium methylate, Sodium methanolate
- CAS Number:124-41-4
- Formula:CH₃NaO
- MDL Number:MFCD00012179
- Molecular Weight:54.02 g/mol
- Merck Index:14,8643
- Melting Pt:>300 °C
- Density:1,3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-699-5
- UN:1431
- ADR:4.2,II
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 124-41-4 Molecular Formula: CH3NaO Formula Weight: 54.02 Melting Point: 300° Beilstein Reference: 3592982 Merck Reference: 14,8643 UN No.: 1431 MDL No.: MFCD00012179
Synonyms: Sodium methylate, Sodium methanolate
- CAS Number:124-41-4
- Formula:CH₃NaO
- MDL Number:MFCD00012179
- Molecular Weight:54.02 g/mol
- Merck Index:14,8643
- Melting Pt:>300 °C
- Density:1,3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-699-5
- UN:1431
- ADR:4.2,II



