

Glutaric anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.20604.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:53 - 57 °C
- Molar Mass:114.1 g/mol
- Bulk Density:990 kg/m3
- Boiling Point:290.5 °C (1013 hPa)
- Vapor Pressure:0.05 hPa (50 °C)
- Flash Point:174 °C
- Density:1.24 g/cm3 (60 °C)
- Ignition Point:320 °C
Specification Test Results
About this item
:
:114.1 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:52…55 °C
Density:1,411 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:174 °C
EINECS:203-593-6
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:108-55-4
Formula:C₅H₆O₃
MDL Number:MFCD00006679