
Bis(2-butoxyethyl)ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-73-2
:
Formula:C₁₂H₂₆O₃
MDL Number:MFCD00009459
:218.34 g/mol
Boiling Pt:256 °C
Melting Pt:–60 °C
Density:0,885
Flash Pt:117 °C (243 °F)
EINECS:204-001-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together