1,3-Diethoxy-2-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4043-59-8
:
Formula:C₇H₁₆O₃
MDL Number:MFCD00026934
Density:0,95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:148.2 g/mol
EINECS:223-736-6
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:148.20 g/mol
- Boiling Point:191 - 192 °C
- Density:0.95 g/cm3 (20 °C)
- Synonyms:Glycerol α,γ-diethyl ether
- Hill Formula:C7H16O3
- Chemical Formula:C2H5OCH2CH(OH)CH2OC2H5
Specification Test Results
Frequently Bought Together