


2-Ethoxyethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-80-5
:MFCD00008590
Formula:C₂H₅OCH₂CH₂OH
MDL Number:MFCD00002869
:90.12 g/mol
Merck Index:13,03786
Boiling Pt:135 °C (1013 hPa)
Melting Pt:–70 °C
Density:0,932 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-804-1
UN:1171
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:40 °C (closed cup)
REACH:01-2119560582-38
:GHS02
:GHS06
:GHS08
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-100 °C
- Molar Mass:90.12 g/mol
- Boiling Point:135 °C (1013 hPa)
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:40 °C
- Refractive Index:1.408 (20 °C, 589 nm)
- Explosion Limit:1.8 - 15.7 %(V)
- Density:0.93 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:235 °C
- Saturation concentration:18 g/m3 (20 °C) Air
- Evaporation number:43
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together