


Diethanolamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-42-2
:
Formula:C₄H₁₁NO₂
MDL Number:MFCD00002843
:105.14 g/mol
Merck Index:14,3107
Boiling Pt:268…270 °C
Melting Pt:27…30 °C
Density:1,097
Flash Pt:176 °C (349 °F)
Storage Temperature:Ambient
EINECS:203-868-0
REACH:01-2119488930-28
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together