Dibutylamine ≥99%, purified
23337.293
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Assay:Min. 99 %
: Di-n-butylamine, N-butyl-1-propanamine
:
:129.25 g/mol
:–62 °C
:203-921-8
:2248
:GHS02
:GHS07
:Ambient
:111-92-2
:C₈H₁₉N
:MFCD00009429
:159
:0.759 g/cm³ (20 °C)
:43
:8 (3) II
:13,03058