1,8-Diaminooctane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:373-44-4
:
Formula:C₈H₂₀N₂
MDL Number:MFCD00008248
:144.26 g/mol
Boiling Pt:225…226 °C
Melting Pt:50…54 °C
Density:0,83 g/cm³ (60 °C)
Flash Pt:106 °C (223 °F)
Storage Temperature:Ambient
EINECS:206-764-3
UN:3259
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together