


1-Propyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:628-30-8
:
Formula:C₄H₇NS
MDL Number:MFCD00004823
:101.17 g/mol
Boiling Pt:152…154 °C
Density:0,977
Flash Pt:46 °C (114 °F)
EINECS:211-035-8
UN:2924
ADR:3,III
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together