

1-Hexyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4404-45-9
:
Formula:C₇H₁₃NS
MDL Number:MFCD00014445
:143.25 g/mol
Boiling Pt:216 °C
Density:0,933
Flash Pt:90 °C (194 °F)
Storage Temperature:Ambient
EINECS:224-549-2
UN:2922
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together