N,N-Dimethylformamide dimethyl acetal ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4637-24-5
:
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482
:119.16 g/mol
Boiling Pt:102…104 °C
Melting Pt:–85 °C
Density:0,897
Flash Pt:7 °C (45 °F)
EINECS:225-063-3
ADR:3,II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together