Order Entry
Northern Ireland
ContactUsLinkComponent
Trimethylsilyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Irritant
Trimethylsilyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
Trimethylsilyl trifluoromethanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    222.26 g/mol
  • Boiling Point:
    35 - 37 °C (13 hPa)
  • Vapor Pressure:
    4.7 hPa (20 °C)
  • Flash Point:
    38 °C DIN 51755 Part 1
  • Density:
    1.23 g/cm3 (20 °C)
  • Ignition Point:
    405 °C DIN 51794

Specification Test Results

Related Information

Frequently Bought Together