Order Entry
Northern Ireland
ContactUsLinkComponent
Trimethylsilyl trifluoromethanesulphonate 99%
Corrosive
Flammable
Trimethylsilyl trifluoromethanesulphonate 99%
Certificates
undefined
Corrosive
Flammable
Trimethylsilyl trifluoromethanesulphonate 99%
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3600 to 1.3620 (20°C, 589 nm)
  • Titration with NaOH:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together