
Trifluoromethanesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:421-83-0
:
Formula:CClF₃O₂S
MDL Number:MFCD00007451
:168.52 g/mol
Boiling Pt:32 °C
Density:1,593
Storage Temperature:Refrigerator
EINECS:207-009-0
UN:3265
ADR:8,II
:GHS05
Related Information
Frequently Bought Together