

Trifluoroacetic anhydride ≥99% (by GC) for synthesis
8.08261.0501
: Merck
:
- Pk:500 mL
- Seq:2725
- Assay (GC, area%):≥99.0% (a/a)
- Identity (IR):Passes Test
- Density (d 20 °C/ 4 °C):1.509 - 1.512
:
:210.03 g/mol
:39,4 °C (1013 hPa)
:–65 °C
:1,487 g/cm³ (20 °C)
:Ambient
:206-982-9
:3265
:8,I
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
:407-25-0
:C₄F₆O₃
:MFCD00000416