
Pimelic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-16-0
:
Formula:C₇H₁₂O₄
MDL Number:MFCD00004425
:160.17 g/mol
Merck Index:14,7431
Boiling Pt:212 °C (10 mmHg)
Melting Pt:102…106 °C
Density:1,329
EINECS:203-840-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together