5-Aminovaleric acid for synthesis
Catalog # 8.41495.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Melting Point:157 - 158 °C (decomposition)
- Molar Mass:117.14 g/mol
Specification Test Results
About this item
:
:117.15 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:157…158 °C
Density:1,33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-544-5
CAS Number:660-88-8
Formula:C₅H₁₁NO₂
MDL Number:MFCD00008232