Order Entry
Northern Ireland
ContactUsLinkComponent
Potassium formate 96% pract.
Irritant
Potassium formate 96% pract.
Certificates
undefined
Irritant
Potassium formate 96% pract.

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    164 to 168 °C
  • Titration with HClO4:
    ≥95.0 %

Specification Test Results

Related Information

Frequently Bought Together