Sodium acetate, anhydrous ≥99.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt: Dec. (on heating)
Melting Pt:324 °C
Density:1,45 g/cm³ (20 °C)
Flash Pt:≥ 250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Specification Test Results
- Assay (HClO₄-titr.):Min. 99.0 %
- Water insoluble matter:Max. 0.005 %
- Loss on drying (120°C):Max. 1.0 %
- pH (5 %, 25 °C):7.0 - 9.2
- Arsenic (As):Max. 0.00001 %
- Aluminium (Al):Max. 0.001 %
- Calcium (Ca):Max. 0.001 %
- Cadmium (Cd):Max. 0.0005 %
- Chromium (Cr):Max. 0.0005 %
- Cobalt (Co):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.02 %
- Magnesium (Mg):Max. 0.0005 %
- Manganese (Mn):Max. 0.0005 %
- Nickel (Ni):Max. 0.0005 %
- Lead (Pb):Max. 0.0005 %
- Zinc (Zn):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.001 %
- Phosphate (PO₄):Max. 0.0005 %
- Sulfate (SO₄):Max. 0.002 %
- Appearance of the solution:complying
- Colour of the solution:complying
- Turbidity of solution:complying
Specification Test Results
Frequently Bought Together