Sodium acetate USP, FCC, J.T.Baker®
3474-07
: Avantor
:
- Pack type:Poly pail, screw top
- Pk:12 kg
- Seq:4187
- Bulk Pharmaceutical Chemical:99.0 - 101.0 %
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤0.035 %
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤0.05 %
- USP - Loss on Drying at 120°C:≤1.0 %
- USP - pH of 3% Solution at 25°C:7.5 - 9.2
- USP - Potassium Compounds:Passes Test
- USP - Residual Acetic Acid, For Information Only:For Information Only
- USP - Sulfate (SO₄):≤0.005 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Alkalinity:≤0.2 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying at 120°C:≤1.0 %
- FCC - Potassium Compounds:Passes Test
- Endotoxin Concentration (2.5 EU/g max.):Passes Test
:
:82.03 g/mol
:12,08711
:881,4 °C (1013 hPa)
:324 °C
:1,528 g/cm³ (20 °C)
:>250 °C (closed cup)
:Ambient
:204-823-8
:01-2119485123-42
:127-09-3
:H₃CCOONa
:MFCD00012459